missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGN46 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | TGN46 |
---|---|
Concentration | 0.33 mg/mL |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16881854
|
Proteintech
13573-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16871854
|
Proteintech
13573-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IASPP is one of the most evolutionarily conserved inhibitors of p53 (TP53; MIM 191170), whereas ASPP1. and ASPP2. are activators of p53.Specifications
TGN46 | |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human | |
O43493 | |
10618 | |
TGN46 Fusion Protein Ag4470 | |
Primary | |
-20°C | |
TGOLN2 |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Tgn38, TGN38 homolog, TGN46, TGN48, TGN51, TGOLN2, trans Golgi network protein 2, TTGN2 | |
TGOLN2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TGN46 Rabbit anti-Human, Polyclonal, Proteintech