Läs mer
Thiabendazole, 98+%, Thermo Scientific™
Fungicide and antihelminthic
Brand: Thermo Scientific Alfa Aesar J60009.36
| Kvantitet | 500g |
|---|
Beskrivning
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
SolubilitySoluble in methanol and dimethyl sulfoxide.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Kemiska identifierare
| C10H7N3S | |
| MFCD00005587 | |
| thiabendazole, tiabendazole, mintezol, equizole, tiabendazol, mintesol, omnizole, thibenzole, thiabendazol, bovizole | |
| CHEBI:45979 | |
| C1=CC=C2C(=C1)NC(=N2)C3=CSC=N3 |
Specifikationer
| Thiabendazole | |
| C10H7N3S | |
| 500g | |
| 611403 | |
| thiabendazole, tiabendazole, mintezol, equizole, tiabendazol, mintesol, omnizole, thibenzole, thiabendazol, bovizole | |
| WJCNZQLZVWNLKY-UHFFFAOYSA-N | |
| 4-(1H-benzimidazol-2-yl)-1,3-thiazole | |
| 5430 | |
| 201.25 |
| 148-79-8 | |
| MFCD00005587 | |
| UN3077 | |
| 14,9289 | |
| Soluble in methanol and dimethyl sulfoxide. | |
| C1=CC=C2C(=C1)NC(=N2)C3=CSC=N3 | |
| 201.247 | |
| CHEBI:45979 | |
| ≥98% |
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.