Learn More
Masterflex™ Thread to Compression Adapters
Compression fittings are an excellent choice for connecting rigid tubing systems
Brand: Masterflex™ 06385-76
This item is not returnable.
View return policy
Description
- Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal
- Available in packs of ten
Specifications
Compression x NPT(M) Connection | |
Polypropylene | |
10 Fittings | |
3/4 in. | |
Ethylene Oxide | |
1 1/4 in. | |
Thread to Compression Adapter |
Not Autoclavable | |
1 13/16 in. | |
3/4 in. | |
NPT(M) 1/2 in. | |
32 mm | |
Straight |
Product Content Correction
Product Title
Masterflex™ Thread to Compression Adapters > 1/2 in.
Spot an opportunity for improvement?Share a Content Correction