missing translation for 'onlineSavingsMsg'
Learn More

THUMPD3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 19807-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16879284

  • 1355.00 SEK / 20µL
Estimated Shipment: 03-04-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

THUMPD3 a protein of unknown function. Its domain structure suggests that it is a methylase. It contains a THUMP domain (an RNA-binding domain) and a UPF0020 domain (putative methylase).
Specifications

Specifications

THUMPD3
Polyclonal
Unconjugated
THUMPD3
THUMP domain containing 3, THUMPD3
Rabbit
Antigen Affinity Chromatography
RUO
14911, 25917, 500288
-20°C
Liquid
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P97770, Q9BV44
Thumpd3
THUMPD3 Fusion Protein Ag13833
20 μL
Primary
Rat, Human, Mouse
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers