Learn More
Thyroglobulin Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21714-1-AP-20UL
This item is not returnable.
View return policy
Description
Thyroglobulin(Tg) is a glycoprotein homodimer produced predomitly by the thryroid gland. It acts as a substrate for the synthesis of thyroxine and triiodothyronine as well as the storage of the inactive forms of thyroid hormone and iodine.Thyroglobulinis secreted from the endoplasmic reticulum to its site of iodination, and subsequent thyroxine biosynthesis, in the follicular lumen. Mutations in this gene cause thyroid dyshormonogenesis, manifested as goiter, and are associated with moderate to severe congenital hypothyroidism. Polymorphisms in this gene are associated with susceptibility to autoimmune thyroid diseases (AITD) such as Graves disease and Hashimoto thryoiditis.Specifications
Thyroglobulin | |
Polyclonal | |
Unconjugated | |
TG | |
AITD3, TG, TGN, thyroglobulin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
7038 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P01266 | |
TG | |
Thyroglobulin Fusion Protein Ag16343 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Product Title
Thyroglobulin Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction