missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIGIT Monoclonal Antibody (GIGD7), Super Bright™ 702, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen 67-9501-80
Additional Details : Weight : 0.01000kg
Description
Rat Monoclonal Antibody
TIGIT Monoclonal antibody specifically detects TIGIT in Mouse samples. It is validated for Flow CytometrySpecifications
TIGIT | |
Monoclonal | |
0.2 mg/mL | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
P86176 | |
TIGIT | |
Affinity chromatography | |
RUO | |
100043314 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
GIGD7 | |
Super Bright 702 | |
TIGIT | |
ENSMUSG00000071552; T-cell immunoreceptor with Ig and ITIM domains; TIGIT; V-set and immunoglobulin domain-containing protein 9; V-set and transmembrane domain-containing protein 3; VSIG9; VSTM3; WUCAM | |
Rat | |
25 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |