Learn More
Tim23 Mouse anti-Human, Unconjugated, Clone: 3H10F5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67535-1-IG-150UL
 View more versions of this product
Product Code. 16890216
  • 4652.90 SEK / 150µL
Explore more special offers
This item is not returnable. View return policy
Description
Description
The Tim23 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Tim23. This antibody recognizes human, mouse, rat antigen. The Tim23 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
Tim23
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, Immunofluorescence
3H10F5
Unconjugated
150 μL
10431
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tim23 Mouse anti-Human, Unconjugated, Clone: 3H10F5, Proteintech > 150 μL