Learn More
Tim23 Mouse anti-Human, Unconjugated, Clone: 3H10F5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67535-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
The Tim23 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Tim23. This antibody recognizes human, mouse, rat antigen. The Tim23 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Tim23 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunohistochemistry, Immunofluorescence | |
3H10F5 | |
Unconjugated | |
20 μL | |
10431 | |
IgG1 |