Läs mer
Tin bis(trifluoromethylsulfonyl)imide, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar 047142.03
| Kvantitet | 1g |
|---|
Beskrivning
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Kemiska identifierare
| 4600096-08-6 | |
| MFCD28100488 | |
| Tin bis(trifluoromethanesulfonyl)imide; Tin bis(trifluoromethanesulfonimide) |
Specifikationer
| Tin bis(trifluoromethylsulfonyl)imide | |
| [(CF3SO2)2N]2Sn | |
| 1g | |
| Hygroscopic | |
| 678.98 |
| 4600096-08-6 | |
| MFCD28100488 | |
| UN3261 | |
| Tin bis(trifluoromethanesulfonyl)imide; Tin bis(trifluoromethanesulfonimide) |