Learn More
Lovibond™ Tintometer™ Calcium Hardness Tablets
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
Brand: Lovibond™ 505191
171.36 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Calcium Hardness Tablets
- Method: Tablet Count
- Bottle of 250
Specifications
Calcium Hardness Test | |
Test kits, colorimeters and spectrophotometers | |
Tablet Count | |
Calcium Hardness Tablets |