Learn More
Sampling Systems™ Tip for Unit Dose Sleeve Sampler
Developed for customers requiring accurate unit dose samples in small volumes. Sampling Systems™ Tip for Unit Dose Sleeve Sampler is ideal for use with a wide range of free flowing powders and granules.
Brand:  Sampling Systems™ 1500W-14
 View more versions of this product
Product Code. 15809281
  • 1125.00 SEK / Each
Only null left
Estimated Shipment: 09-09-2025
Description
  • For sampling small volume powders and granules.
  • Chemically resistant 316L stainless steel construction.
  • Different sample sizes can be taken using the same sampler
  • Tips for use with Type 1 or Type 2 Sampling Chambers
Specifications
Specifications
Unit Dose Sleeve Sampler Tip
1.4 mL
31.5 mm
Stainless Steel (316L)
8 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Sampling Systems™ Tip for Unit Dose Sleeve Sampler > Capacity: 1.4mL