missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIP30 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10251-1-AP-150UL
This item is not returnable.
View return policy
Description
CC3 (HTATIP2) is a member of the short-chain dehydrogenases/reductases (SDR) family. It is a novel serine/threonine kinase that phosphorylates the C-terminal domain (CTD) of the largest RNA polymerase II subunit and induces the expression of apoptosis related genes Bad and Siva, as well as metastasis suppressor NM23-H2. It also interacts with an estrogen receptor alpha-interacting coactivator CIA and regulates ERalpha-mediated c-myc transcription.Specifications
TIP30 | |
Polyclonal | |
Unconjugated | |
HTATIP2 | |
CC3, HTATIP2, Oxidoreductase HTATIP2, SDR44U1, TIP30 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10553, 292935, 53415 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9BUP3, Q9Z2G9 | |
Htatip2 | |
TIP30 Fusion Protein Ag0353 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TIP30 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction