missing translation for 'onlineSavingsMsg'
Learn More
Titanium(Iv) Chloride Solution, 1.0M in methylene chloride, Honeywell™ Product Code.: 15662710

Titanium(Iv) Chloride Solution, 1.0M in methylene chloride, Honeywell™

Product Code. 15662710 Shop All Honeywell Fluka Products
800 mL, Glass Bottle, 800mL
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
800 mL
Packaging:
Glass Bottle
missing translation for 'unitSize'
100mL
800mL
This item is not returnable. View return policy

Product Code. 15662710

Brand: Honeywell-Fluka 249866800ML

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

1.0 M in methylene chloride

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7550-45-0
Molecular Formula Cl4Ti
Molecular Weight (g/mol) 189.667
MDL Number MFCD00011267
InChI Key XJDNKRIXUMDJCW-UHFFFAOYSA-J
Synonym titanium iv chloride, titanium 4+ tetrachloride, acmc-1bjgv, titanium 4+ ion tetrachloride, parent, titanium +4 cation tetrachloride, titanium iv chloride 100ml, superlist names titanium chloride, t-4, titanic chloride; titanio tetracloruro di, unii-8o3pje5t7q; titanium chloride ticl4
PubChem CID 160960
IUPAC Name titanium(4+);tetrachloride
SMILES [Cl-].[Cl-].[Cl-].[Cl-].[Ti+4]

Specifications

Quantity 800 mL
Linear Formula TiCl4
UN Number UN3289
Formula Weight 189.68g/mol
Concentration or Composition (by Analyte or Components) 1.0M in methylene chloride
Packaging Glass Bottle
Vapor Pressure 24.09psi (55°C), 6.93psi (20°C)
Density 1.365g/mL at 25°C
Chemical Name or Material Titanium(IV) Chloride Solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Titanium(Iv) Chloride Solution, 1.0M in methylene chloride, Honeywell™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.