missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | TK1 |
---|---|
Concentration | 0.19 mg/mL |
Applications | Western Blot, Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16894974
|
Proteintech
15691-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16884974
|
Proteintech
15691-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Thymidine Kinase (TK1) is a highly conserved phosphotransferase that is present in most living cells. Thymidine Kinase catalyzes the phosphorylation reaction: deoxythymidine + ATP = deoxythymidine 5′-phosphate + ADP; it is thus involved in the reaction chain to introduce deoxythymidine into the DNA. Thymidine kinase is required for the action of many antiviral drugs, such as azidothymidine (AZT), and is also used to select hybridoma cell lines in the production of monoclonal antibodies. Thymidine Kinase has many clinical applications as it is only present in anticipation of cell division.Specifications
TK1 | |
Western Blot, Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
P04183, P04184, P27158 | |
21877, 24834, 7083 | |
TK1 Fusion Protein Ag8249 | |
Primary | |
-20°C | |
TK1 |
0.19 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
thymidine kinase 1, soluble, Thymidine kinase, cytosolic, TK1, TK2 | |
TK1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech