missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLE1 Mouse anti-Human, Clone: 2F2H11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66216-1-IG-150UL
This item is not returnable.
View return policy
Description
Transcriptional corepressor that binds to a number of transcription factors. Inhibits NF-kappa-B-regulated gene expression. Inhibits the transcriptional activation mediated by FOXA2, and by CTNNB1 and TCF family members in Wnt signaling. The effects of full-length TLE family members may be modulated by association with domit-negative AES. Unusual function as coactivator for ESRRG.Specifications
TLE1 | |
Monoclonal | |
1.05 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q04724 | |
TLE1 | |
TLE1 Fusion Protein Ag21327 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunohistochemistry (Paraffin), Western Blot | |
2F2H11 | |
Unconjugated | |
TLE1 | |
E(Sp1) homolog, ESG, ESG1, GRG1, TLE1 | |
Mouse | |
Protein A | |
RUO | |
7088 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TLE1 Mouse anti-Human, Clone: 2F2H11, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction