missing translation for 'onlineSavingsMsg'
Learn More

TLR5 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 4B3D2, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66570-1-IG-20UL

 View more versions of this product

Product Code. 16888565

  • 1645.54 SEK / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TLR5 is a Type I membrane protein belonging to the Toll-like receptor family, participates in the innate immune response to microbial agents. TLR5 also plays a role in mediating detection of bacterial flagellins. TLR5 acts via MyD88 and TRAF6, leading to NF-kappa-B activation, cytokine secretion and the inflammatory response. TLR5 protein binds to TIRAP and MyD88 via their respective TIR domains TLR5 is highly expressed in liver, and is detected in lung and at very low levels in most other tissues. The TLR5 gene lies in a locus that is associated with susceptibility to Salmonella. Inbred strains of mice can be classified into 3 categories according to their resistance to infection with S.typhimurium: susceptible (BALB/c, C57BL/6, C3H/He), intermediate (DBA/2, C75L) and resistant (A, CBA). The strain MOLF/Ei is highly susceptible to the infection, has a unique TLR5 haplotype and a lower expression of TRL5. A common TLR5 stop codon polymorphism that disrupts TLR5 signaling is associated with susceptibility to Legionnaires′ disease and demonstrates the importance of TLR5 in the innate immune response.
Specifications

Specifications

TLR5
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O60602, Q9JLF7
Tlr5
TLR5 Fusion Protein Ag13840
20 ÎĽL
Primary
Pig, Human, Mouse, Rat
Antibody
IgG1
Western Blot
4B3D2
Unconjugated
TLR5
SLEB1, TIL3, TLR5, toll like receptor 5
Mouse
Protein G
RUO
100144476, 289337, 53791, 7100
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
TLR5 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 4B3D2, Proteintech > 20 ÎĽL; Unconjugated

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.