missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TM7SF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | TM7SF2 |
---|---|
Concentration | 0.33 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16879503
|
Proteintech
12033-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16869503
|
Proteintech
12033-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TM7SF2 is involved in the conversion of lanosterol to cholesterol.Specifications
TM7SF2 | |
Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
O76062, Q71KT5 | |
293688, 7108, 73166 | |
TM7SF2 Fusion Protein Ag2637 | |
Primary | |
-20°C | |
TM7SF2 |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Another new gene 1 protein, C 14 sterol reductase, Delta 14 SR, Delta(14) sterol reductase, DHCR14A, NET47, Putative sterol reductase SR 1, Sterol C14 reductase, TM7SF2 | |
TM7SF2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TM7SF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech