Learn More
TM9SF4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25595-1-AP-20UL
 View more versions of this product
Product Code. 16884155
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
TM9SF4 is a protein coding gene.
Specifications
Specifications
TM9SF4
Polyclonal
Unconjugated
TM9SF4
KIAA0255, TM9SF4
Rabbit
Antigen Affinity Chromatography
RUO
9777
-20°C
Liquid
Western Blot
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q92544
TM9SF4
TM9SF4 Fusion Protein Ag22335
20 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TM9SF4 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated