missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMCO6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20117-1-AP-20UL
This item is not returnable.
View return policy
Specifications
TMCO6 | |
Polyclonal | |
Unconjugated | |
TMCO6 | |
PRO1580, TMCO6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
291661, 55374, 71983 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8BQX5, Q96DC7 | |
Tmco6 | |
TMCO6 Fusion Protein Ag13992 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TMCO6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction