missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMED3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | TMED3 |
---|---|
Concentration | 0.4 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16891445
|
Proteintech
21902-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16881445
|
Proteintech
21902-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The function remains known.Specifications
TMED3 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
Q6AY25, Q78IS1, Q9Y3Q3 | |
23423, 300888, 66111 | |
TMED3 Fusion Protein Ag14032 | |
Primary | |
-20°C | |
TMED3 |
0.4 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
C15orf22, Membrane protein p24B, P24B, TMED3 | |
Tmed3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TMED3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech