Learn More
TMEM11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16564-1-AP-20UL
 View more versions of this product
Product Code. 16896214
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
Plays a role in mitochondrial morphogenesis.
Specifications
Specifications
TMEM11
Polyclonal
Unconjugated
TMEM11
C17orf35, PM1, PMI, Protein PM1, TMEM11, transmembrane protein 11
Rabbit
Antigen Affinity Chromatography
RUO
216821, 303196, 8834
-20°C
Liquid
Western Blot
0.24 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
B0BN86, P17152, Q8BK08
TMEM11
TMEM11 Fusion Protein Ag9839
20 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TMEM11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated