Learn More
TMEM111 Mouse anti-Human, Unconjugated, Clone: 2F8G5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67205-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
The TMEM111 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TMEM111. This antibody recognizes human, mouse, rat, pig antigen. The TMEM111 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
TMEM111 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Mouse, Rat, Pig |
Western Blot, Immunoprecipitation, Immunohistochemistry, ELISA | |
2F8G5 | |
Unconjugated | |
20 μL | |
55831 | |
IgG1 |