Learn More
TMEM111 Mouse anti-Human, Unconjugated, Clone: 2F8G5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67205-1-IG-150UL
 View more versions of this product
Product Code. 16879715
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
The TMEM111 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TMEM111. This antibody recognizes human, mouse, rat, pig antigen. The TMEM111 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
TMEM111
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, Immunoprecipitation, Immunohistochemistry, ELISA
2F8G5
Unconjugated
150 μL
55831
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TMEM111 Mouse anti-Human, Unconjugated, Clone: 2F8G5, Proteintech > 150 μL