missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM179 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24799-1-AP-20UL
This item is not returnable.
View return policy
Specifications
TMEM179 | |
Polyclonal | |
Unconjugated | |
TMEM179 | |
C14orf90, TMEM179, TMEM179A, transmembrane protein 179, Transmembrane protein 179A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
104885, 388021 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6ZVK1, Q8BHH9 | |
TMEM179 | |
TMEM179 Fusion Protein Ag20502 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TMEM179 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction