missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM201 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24092-1-AP-150UL
Additional Details : Weight : 0.00010kg
Specifications
TMEM201 | |
Polyclonal | |
Unconjugated | |
TMEM201 | |
NET5, RP13 15M17.2, SAMP1, TMEM201, transmembrane protein 201 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
199953, 230917 | |
-20°C | |
Liquid |
Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A2A8U2, Q5SNT2 | |
TMEM201 | |
TMEM201 Fusion Protein Ag21360 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |