missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM201 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | TMEM201 |
---|---|
Concentration | 0.17 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16882665
|
Proteintech
24092-1-AP-20UL |
20 μL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16872665
|
Proteintech
24092-1-AP-150UL |
150 μL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
TMEM201 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
A2A8U2, Q5SNT2 | |
199953, 230917 | |
TMEM201 Fusion Protein Ag21360 | |
Primary | |
-20°C | |
TMEM201 |
0.17 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
NET5, RP13 15M17.2, SAMP1, TMEM201, transmembrane protein 201 | |
TMEM201 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TMEM201 Rabbit anti-Human, Mouse, Polyclonal, Proteintech