missing translation for 'onlineSavingsMsg'
Learn More

TMEM201 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 24092-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16882665

  • 1656.73 SEK / 20µL
Estimated Shipment: 14-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

TMEM201
Polyclonal
Unconjugated
TMEM201
NET5, RP13 15M17.2, SAMP1, TMEM201, transmembrane protein 201
Rabbit
Antigen Affinity Chromatography
RUO
199953, 230917
-20°C
Liquid
Western Blot
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
A2A8U2, Q5SNT2
TMEM201
TMEM201 Fusion Protein Ag21360
20 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers