missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM61 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20997-1-AP-150UL
This item is not returnable.
View return policy
Specifications
TMEM61 | |
Polyclonal | |
Unconjugated | |
TMEM61 | |
TMEM61, transmembrane protein 61 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
199964 | |
-20°C | |
Liquid |
Western Blot | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8N0U2 | |
TMEM61 | |
TMEM61 Fusion Protein Ag15044 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TMEM61 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction