Learn More
TMEM70 Mouse anti-Human, Mouse, Clone: 2C11C3, Proteintech
Mouse Monoclonal Antibody
1662.46 SEK - 4700.76 SEK
Specifications
Antigen | TMEM70 |
---|---|
Clone | 2C11C3 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16887185
|
Proteintech
60195-1-IG-20UL |
20 μL |
1662.46 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16877185
|
Proteintech
60195-1-IG-150UL |
150 μL |
4700.76 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TMEM70 is a recently identified mitochondrial protein that is thought to play a role in the biogenesis of the ATP synthase in higher eukaryotes. Mutations in this gene result in early neonatal onset of hypotonia, hypertrophic cardiomyopathy, lactic acidos and 3-methylglutaconic aciduria (3-MGC-uria), and usually cause death within the first six weeks of life, although some patients survive much longer. Little is known of the role of TMEM70, but it is conserved across multicellular eukaryotic organisms. It contains a conserved DUF1301 domain and two putative transmembrane regions.Specifications
TMEM70 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
TMEM70, transmembrane protein 70 | |
TMEM70 | |
IgG2b | |
Protein A | |
Antibody |
2C11C3 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Mouse, Human | |
Q921N7, Q9BUB7 | |
54968, 70397 | |
TMEM70 Fusion Protein Ag16947 | |
Primary | |
-20°C | |
TMEM70 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
TMEM70 Mouse anti-Human, Mouse, Clone: 2C11C3, Proteintech