missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNF beta Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13111-1-AP-20UL
This item is not returnable.
View return policy
Description
TNF beta, a member of the tumor necrosis factor family, is a cytokine produced by lymphocytes. The protein is highly inducible, secreted, and forms heterotrimers with lymphotoxin-beta which anchor lymphotoxin-alpha to the cell surface. This protein also mediates a large variety of inflammatory, immunostimulatory, and antiviral responses, is involved in the formation of secondary lymphoid organs during development and plays a role in apoptosis. Genetic variations in this gene are associated with susceptibility to leprosy type 4 and psoriatic arthritis.Specifications
TNF beta | |
Polyclonal | |
Unconjugated | |
LTA | |
LT, LT alpha, LTA, Lymphotoxin alpha, TNF beta, TNFB, TNFSF1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4049 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P01374 | |
LTA | |
TNF beta Fusion Protein Ag3775 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TNF beta Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction