missing translation for 'onlineSavingsMsg'
Learn More
Tobramycin, MP Biomedicals™ Product Code.: 11485321

Tobramycin, MP Biomedicals™

Product Code. 11485321 Shop All MP Biomedical Products
0.1 g, 100mg
Click to view available options
Quantity:
0.1 g
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 11485321

Brand: MP Biomedicals 0215692680

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 32986-56-4
Molecular Formula C18H37N5O9
Molecular Weight (g/mol) 467.52
InChI Key NLVFBUXFDBBNBW-PBSUHMDJSA-N
Synonym Distobram, Gernebcin
PubChem CID 36294
ChEBI CHEBI:28864
IUPAC Name (2S,3R,4S,5S,6R)-4-amino-2-[(1S,2S,3R,4S,6R)-4,6-diamino-3-[(2R,3R,5S,6R)-3-amino-6-(aminomethyl)-5-hydroxyoxan-2-yl]oxy-2-hydroxycyclohexyl]oxy-6-(hydroxymethyl)oxane-3,5-diol
SMILES C1C(C(C(C(C1N)OC2C(C(C(C(O2)CO)O)N)O)O)OC3C(CC(C(O3)CN)O)N)N

Specifications

Type Tobramycin
Physical Form Powder
Color White
Quantity 0.1 g
Formula Weight 467.5
Chemical Name or Material Tobramycin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tobramycin, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.