missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOP1MT Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16540-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a mitochondrial DNA topoisomerase that plays a role in the modification of DNA topology. The encoded protein is a type IB topoisomerase and catalyzes the transient breaking and rejoining of DNA to relieve tension and DNA supercoiling generated in the mitochondrial genome during replication and transcription. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.Specifications
TOP1MT | |
Polyclonal | |
Unconjugated | |
TOP1MT | |
2900052H09Rik, TOP1MT | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
116447 | |
-20°C | |
Liquid |
Western Blot | |
0.44 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q969P6 | |
TOP1MT | |
TOP1MT Fusion Protein Ag9857 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TOP1MT Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction