missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOR1AIP2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24769-1-AP-20UL
This item is not returnable.
View return policy
Description
One of the two protein isoforms encoded by this gene is a type II integral membrane protein found in the endoplasmic reticulum (ER). The encoded protein is a cofactor for the ATPase TorsinA, regulating the amount of TorsinA present in the ER compared to that found in the nuclear envelope. Defects in this protein are a cause of early onset primary dystonia, a neuromuscular disease. The other isoform encoded by this gene is an interferon alpha responsive protein whose cellular role has yet to be determined.Specifications
TOR1AIP2 | |
Polyclonal | |
Unconjugated | |
TOR1AIP2 | |
LULL1, Lumenal domain like LAP1, RP11 12M5.5, TOR1AIP2, torsin A interacting protein 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
163590 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunocytochemistry | |
0.49 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9H496 | |
TOR1AIP2 | |
TOR1AIP2 Fusion Protein Ag20126 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TOR1AIP2 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction