missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPPII Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14120-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a ubiquitously expressed transcription factor that controls cholesterol homeostasis by stimulating transcription of sterol-regulated genes. The encoded protein contains a basic helix-loop-helix-leucine zipper (bHLH-Zip) domain.Specifications
TPPII | |
Polyclonal | |
Unconjugated | |
TPP2 | |
TPP 2, TPP II, TPP2, TPPII, Tripeptidyl aminopeptidase, Tripeptidyl peptidase 2, Tripeptidyl peptidase II | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22019, 7174 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Flow Cytometry, Immunoprecipitation | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P29144, Q64514 | |
TPP2 | |
TPPII Fusion Protein Ag5296 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |