missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPPII Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | TPPII |
---|---|
Concentration | 0.3 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Flow Cytometry, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16892614
|
Proteintech
14120-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16882614
|
Proteintech
14120-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a ubiquitously expressed transcription factor that controls cholesterol homeostasis by stimulating transcription of sterol-regulated genes. The encoded protein contains a basic helix-loop-helix-leucine zipper (bHLH-Zip) domain.Specifications
TPPII | |
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Flow Cytometry, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
P29144, Q64514 | |
22019, 7174 | |
TPPII Fusion Protein Ag5296 | |
Primary | |
-20°C | |
TPP2 |
0.3 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
TPP 2, TPP II, TPP2, TPPII, Tripeptidyl aminopeptidase, Tripeptidyl peptidase 2, Tripeptidyl peptidase II | |
TPP2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TPPII Rabbit anti-Human, Mouse, Polyclonal, Proteintech