missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPPP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | TPPP2 |
---|---|
Concentration | 0.18 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16881944
|
Proteintech
13633-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16871944
|
Proteintech
13633-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
TPPP2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
P59282, Q0P5Y3 | |
122664, 219038, 498497 | |
TPPP2 Fusion Protein Ag4561 | |
Primary | |
-20°C | |
TPPP2 |
0.18 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
C14orf8, P18, p25beta, Protein p25 beta, TPPP/p18, TPPP2 | |
Tppp2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TPPP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech