missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPSAB1 Mouse anti-Human, Clone: 6A10G1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66174-1-IG-20UL
This item is not returnable.
View return policy
Description
Tryptases comprise a family of trypsin-like serine proteases, the peptidase family S1. Tryptases are enzymatically active only as heparin-stabilized tetramers, and they are resistant to all known endogenous proteinase inhibitors. Several tryptase genes are clustered on chromosome 16p13.3. These genes are characterized by several distinct features. They have a highly conserved 3′ UTR and contain tandem repeat sequences at the 5′ flank and 3′ UTR which are thought to play a role in regulation of the mRNA stability. These genes have an intron immediately upstream of the initiator Met codon, which separates the site of transcription initiation from protein coding sequence. This feature is characteristic of tryptases but is unusual in other genes. The alleles of this gene exhibit an unusual amount of sequence variation, such that the alleles were once thought to represent two separate genes, alpha and beta 1. Beta tryptases appear to be the main isoenzymes expressed in mast cells, whereas in basophils, alpha tryptases predominate. Tryptases have been implicated as mediators in the pathogenesis of asthma and other allergic and inflammatory disorders.Specifications
TPSAB1 | |
Monoclonal | |
0.69 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q15661 | |
TPSAB1 | |
TPSAB1 Fusion Protein Ag4167 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot, Immunohistochemistry (Paraffin) | |
6A10G1 | |
Unconjugated | |
TPSAB1 | |
MCP7, TPS1, TPS2, TPSAB1, TPSB1, Tryptase 1, tryptase alpha/beta 1, Tryptase beta 1, Tryptase I | |
Mouse | |
Protein G | |
RUO | |
7177 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TPSAB1 Mouse anti-Human, Clone: 6A10G1, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction