Learn More
TPST1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | TPST1 |
---|---|
Concentration | 233 ÎĽg/mL |
Applications | Western Blot, Immunohistochemistry, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16890104
|
Proteintech
12404-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16880104
|
Proteintech
12404-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The TPST1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TPST1. This antibody recognizes human, mouse, rat antigen. The TPST1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
TPST1 | |
Western Blot, Immunohistochemistry, ELISA | |
Unconjugated | |
Human, Rat, Mouse | |
IgG |
233 ÎĽg/mL | |
Polyclonal | |
Rabbit | |
8460 | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.