Learn More
TRAF5 Mouse anti-Human, Unconjugated, Clone: 1B1A7, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66991-1-IG-20UL
 View more versions of this product
Product Code. 16889335
  • 1645.54 SEK / 20µL
Description
The TRAF5 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TRAF5. This antibody recognizes human antigen. The TRAF5 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
TRAF5
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, ELISA
1B1A7
Unconjugated
20 μL
7188
IgG1
Certificates
Promotions
Promotions
Product Content Correction
Product Title
TRAF5 Mouse anti-Human, Unconjugated, Clone: 1B1A7, Proteintech > 20 μL