Learn More
TRAF5 Mouse anti-Human, Unconjugated, Clone: 1B1A7, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66991-1-IG-150UL
 View more versions of this product
Product Code. 16879335
  • 4652.90 SEK / 150µL
Description
The TRAF5 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TRAF5. This antibody recognizes human antigen. The TRAF5 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
TRAF5
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, ELISA
1B1A7
Unconjugated
150 μL
7188
IgG1
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
TRAF5 Mouse anti-Human, Unconjugated, Clone: 1B1A7, Proteintech > 150 μL