Learn More
TRAF5 Mouse anti-Human, Unconjugated, Clone: 1B1A7, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66991-1-IG-20UL
 View more versions of this product
Product Code. 16889335
  • 1645.54 SEK / 20µL
Explore more special offers
This item is not returnable. View return policy
Description
Description
The TRAF5 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TRAF5. This antibody recognizes human antigen. The TRAF5 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
TRAF5
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, ELISA
1B1A7
Unconjugated
20 μL
7188
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TRAF5 Mouse anti-Human, Unconjugated, Clone: 1B1A7, Proteintech > 20 μL