Läs mer
[trans-4-(aminomethyl)cyclohexyl]methanol, TRC
Produktkod. 30456769
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30456769 500 mg
30455600 100 mg
30450320 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30456769 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT312396500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 17879-23-1
Molekylformel C8H17NO
Molekylvikt (g/mol) 143.22
IUPAC-namn [4-(aminomethyl)cyclohexyl]methanol
LEDER NC[C@@H]1CC[C@@H](CO)CC1
Specifikationer
Kemiskt namn eller material [trans-4-(aminomethyl)cyclohexyl]methanol
Kvantitet 500 mg
InChI formel InChI=1S/C8H17NO/c9-5-7-1-3-8(6-10)4-2-7/h7-8,10H,1-6,9H2/t7-,8-
Formel vikt 143.131