missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transgelin 2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10234-2-AP-150UL
This item is not returnable.
View return policy
Description
This antibody was generated against full length transgenlin 2 protein. It may cross react with other two transgelins based on the sequence similarity.
The protein encoded by this gene is a homolog of the protein transgelin, which is one of the earliest markers of differentiated smooth muscle. The function of this protein has not yet been determined.Specifications
Transgelin 2 | |
Polyclonal | |
Unconjugated | |
TAGLN2 | |
SM22 beta, SM22beta, TAGLN2, Transgelin 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21346, 8407 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P37802, Q9WVA4 | |
TAGLN2 | |
Transgelin 2 Fusion Protein Ag0337 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Transgelin 2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction