missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trappc9/NIBP Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1355.00 SEK - 3665.00 SEK
Specifications
Antigen | Trappc9/NIBP |
---|---|
Concentration | 0.31 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16829814
|
Proteintech
20287-1-AP-150UL |
150 μL |
3665.00 SEK
150µL |
Estimated Shipment: 07-02-2023 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
16839814
|
Proteintech
20287-1-AP-20UL |
20 μL |
1355.00 SEK
20µL |
Estimated Shipment: 07-02-2023 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
Description
NIBP is a 1246 amino acid intracellular regulatory protein. Reports suggest that NIBP physically interacts with NIK, IKK (beta), but not IKK (alpha) or IKK (gamma) and thus acts as a novel enhancer of cytokine-induced NF-(kappa) B signaling pathway. Due to its neuronal expression NIBP is considered as a potential target for modulating the NF-(kappa) B signaling cascade in neuronal pathologies dependent upon abnormal activation of this pathway. NIBP is mainly expressed in brain, muscle, heart, and kidney.Specifications
Trappc9/NIBP | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q3U0M1, Q96Q05 | |
76510, 83696 | |
Trappc9,NIBP Fusion Protein Ag14017 | |
Primary | |
-20°C | |
TRAPPC9 |
0.31 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
1810044A24Rik, 2900005P22Rik, 4632408O18Rik, Ibp, KIAA1882, mKIAA1882, NIBP, Trappc9, TRS130 | |
Trappc9 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |