Learn More
TREML4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 18056-1-AP-150UL
 View more versions of this product
Product Code. 16888094
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
Positively regulates Toll-like receptor TLR7 signaling in macrophages.
Specifications
Specifications
TREML4
Polyclonal
Unconjugated
TREML4
TLT 4, TLT4, Trem like transcript 4 protein, TREML4
Rabbit
Antigen Affinity Chromatography
RUO
224840, 285852
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.4 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q3LRV9, Q6UXN2
TREML4
TREML4 Fusion Protein Ag12574
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TREML4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated