Learn More
Tri-n-octylphosphine, tech. 90%
Description
Tri-n-octylphosphine is used for coating zinc sulfide shells on cadmium-selenium quantum dot core by successive ionic layer adsorption and reaction method. It acts as a precursor to trioctylphosphine oxide. Further, it is used as a solvent for cadmium and selenium precursors. It also serves as a common reagent in the chemical synthesis of nanoparticles. It acts as a precursor to trioctylphosphine oxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tri-n-octylphosphine is used for coating zinc sulfide shells on cadmium-selenium quantum dot core by successive ionic layer adsorption and reaction method. It acts as a precursor to trioctylphosphine oxide. Further, it is used as a solvent for cadmium and selenium precursors. It also serves as a common reagent in the chemical synthesis of nanoparticles. It acts as a precursor to trioctylphosphine oxide.
Solubility
Immiscible with water.
Notes
Air sensitive. Incompatible with strong oxidizing agents.

Specifications
Specifications
Density | 0.831 |
Boiling Point | 240°C (10 mmHg) |
Flash Point | 147°C (296°F) |
Odor | Garlic-like |
Refractive Index | 1.468 |
Quantity | 25 mL |
UN Number | UN1760 |
Beilstein | 1776995 |
Sensitivity | Air Sensitive |
Solubility Information | Immiscible with water. |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
10241624 | Certificate of Analysis | 12/04/2024 | L15744.AP,L15744.AC,L15744.AE |
10207192 | Certificate of Analysis | 08/12/2021 | L15744.AP,L15744.AC,L15744.AE |
10196841 | Certificate of Analysis | 08/12/2021 | L15744.AP,L15744.AC,L15744.AE |
5003A21R | Certificate of Analysis | 08/12/2021 | L15744.AP,L15744.AC,L15744.AE |
10201172 | Certificate of Analysis | 08/12/2021 | L15744.AP,L15744.AC,L15744.AE |
Safety and Handling

- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.