Learn More
TRIM9 Mouse anti-Human, Unconjugated, Clone: 1D2B10, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67268-1-IG-150UL
 View more versions of this product
Product Code. 16879795
  • 4652.90 SEK / 150µL
Description
The TRIM9 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TRIM9. This antibody recognizes human, mouse, rat, pig antigen. The TRIM9 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
TRIM9
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, Immunohistochemistry, ELISA
1D2B10
Unconjugated
150 μL
114088
IgG1
Certificates
Promotions
Promotions
Product Content Correction
Product Title
TRIM9 Mouse anti-Human, Unconjugated, Clone: 1D2B10, Proteintech > 150 μL