missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIP4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12324-1-AP-20UL
This item is not returnable.
View return policy
Description
Transcription coactivator of nuclear receptors which functions in conjunction with CBP-p300 and SRC-1 and may play an important role in establishing distinct coactivator complexes under different cellular conditions. Plays a pivotal role in the transactivation of NF-kappa-B, SRF and AP1. Acts as a mediator of transrepression between nuclear receptor and either AP1 or NF-kappa-B. Plays a role in androgen receptor transactivation and in testicular function.Specifications
TRIP4 | |
Polyclonal | |
Unconjugated | |
TRIP4 | |
ASC 1, HsT17391, TR interacting protein 4, TRIP 4, TRIP4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
315769, 56404, 9325 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q15650, Q9QXN3 | |
Trip4 | |
TRIP4 Fusion Protein Ag2987 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TRIP4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction