Learn More
TRMT5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18255-1-AP-150UL
This item is not returnable.
View return policy
Description
tRNAs contain as many as 13 or 14 nucleotides that are modified posttranscriptionally by enzymes that are highly specific for particular nucleotides in the tRNA structure. TRMT5 methylates the N1 position of guanosine-37 in selected tRNAs using S-adenosyl methionine.Specifications
TRMT5 | |
Polyclonal | |
Unconjugated | |
TRMT5 | |
KIAA1393, M1G methyltransferase, TRM5, TRMT5, tRNA [GM37] methyltransferase, tRNA methyltransferase 5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
362754, 57570, 76357 | |
-20°C | |
Liquid |
Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q32P41, Q9D0C4 | |
Trmt5 | |
TRMT5 Fusion Protein Ag13075 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Product Title
TRMT5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction