missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPC4AP Mouse anti-Human, Clone: 2C10B1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66138-1-IG-150UL
This item is not returnable.
View return policy
Description
Substrate-specific adapter of a DCX (DDB1-CUL4-X-box) E3 ubiquitin-protein ligase complex required for cell cycle control. The DCX(TRUSS) complex specifically mediates the polyubiquitination and subsequent degradation of MYC. Also participates in the activation of NFKB1 in response to ligation of TNFRSF1A, possibly by linking TNFRSF1A to the IKK signalosome. Involved in JNK activation via its interaction with TRAF2. Also involved in elevation of endoplasmic reticulum Ca(2+) storage reduction in response to CHRM1.Specifications
TRPC4AP | |
Monoclonal | |
0.96 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8TEL6 | |
TRPC4AP | |
TRPC4AP Fusion Protein Ag18772 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG3 |
Immunohistochemistry (Paraffin), Western Blot | |
2C10B1 | |
Unconjugated | |
TRPC4AP | |
C20orf188, Protein TAP1, Protein TRUSS, Trp4 associated protein, Trpc4 associated protein, TRPC4AP, TRRP4AP, TRUSS | |
Mouse | |
Protein A | |
RUO | |
26133 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TRPC4AP Mouse anti-Human, Clone: 2C10B1, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction